missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium Powder, -60 mesh, 99.99% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4455622

 View more versions of this product

Catalog No. AA4455622



Specifications

Specifications

Chromium
7440-47-3
99.99% (Metals basis)
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
[Cr]
51.996
CHEBI:28073
Gray
100g
Gray powder
Cr
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
Odorless
chromium
23976
Powder

Chemical Identifiers

7440-47-3
51.996
VYZAMTAEIAYCRO-UHFFFAOYSA-N
23976
chromium
Cr
MFCD00010944
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
CHEBI:28073
[Cr]
Safety and Handling

Safety and Handling

H351; Suspected of causing cancer.

EINECSNumber : 231-157-5

SDS Documents