missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium Powder, -325 mesh, 99% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1014836

 View more versions of this product

Catalog No. AA1014836



Specifications

Specifications

Chromium
7440-47-3
99% (Metals basis)
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chromium
51.996
CHEBI:28073
Gray
500g
Gray powder
Cr
ccris 159
[Cr]
Odorless
23976
Powder

Chemical Identifiers

7440-47-3
51.996
VYZAMTAEIAYCRO-UHFFFAOYSA-N
23976
chromium
Cr
MFCD00010944
ccris 159
CHEBI:28073
[Cr]
Safety and Handling

Safety and Handling

H228-H351; Flammable solid. Suspected of causing cancer.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-157-5

Documents