Please login to your online account to display your discounted pricing

Chromium powder, -200 mesh, 99.95% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Chromium powder
CAS 7440-47-3
MDL Number MFCD00010944
UN Number UN3089
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1379614

  SDS
alfa aesar
1379614
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 25g
Chemical Name or Material Chromium powder
TSCA TSCA
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00010944
CAS 7440-47-3
UN Number UN3089
Mesh Size −200 mesh
Odor Odorless
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228-H351;
Flammable solid.
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P210-P280-P281-P308+P313-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Silver
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
RTECS Number GB4200000
Safety and Handling

Warning

GHS P Statement
P210-P280-P281-P308+P313-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H351;
Flammable solid.
Suspected of causing cancer.