Learn More
Chromium powder, -200 mesh, 99.94% (metals basis), Thermo Scientific Chemicals
Chromium, Cr, CAS Number-7440-47-3, chrome, chromium, elemental, ccris 159, chrome french, compounds, chrom, chrom german, cromo, metal, chromium, metal, 500g, Gray powder, 99.95% (Metals basis), CHEBI:28073, DOT Class: 4.1, Packaging Group: III, 231-157-5, H228
Supplier: Thermo Scientific Chemicals 01379636
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
7440-47-3 | |
99.95% (Metals basis) | |
MFCD00010944 | |
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
[Cr] | |
52.00 | |
CHEBI:28073 | |
Odorless |
Chromium | |
Gray powder | |
Cr | |
500 g | |
VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
chromium | |
23976 | |
-200 Mesh | |
Powder |
Chemical Identifiers
7440-47-3 | |
52.00 | |
VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
23976 | |
chromium |
Cr | |
MFCD00010944 | |
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
CHEBI:28073 | |
[Cr] |
Safety and Handling
Wear suitable protective clothing and gloves.
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-157-5
RTECSNumber : GB4200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only