missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium plasma standard solution, Alfa Aesar™, Specpure™

Chemical Identifiers

CAS 7647-01-0,7440-47-3,7732-18-5
Physical Form Liquid
Molecular Formula Cr in 5% HCl
MDL Number MFCD00010944
Chemical Name or Material Chromium
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA13864AD SDS Alfa Aesar
13864AD
50mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA13864AE SDS Alfa Aesar
13864AE
100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA13864AP SDS Alfa Aesar
13864AP
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7647-01-0,7440-47-3,7732-18-5
Cr in 5% HCl
Chromium
Liquid
MFCD00010944

Specifications

7647-01-0,7440-47-3,7732-18-5
Liquid
MFCD00010944
23 hPa (17mm Hg) at 20°C
50mL
Clear liquid
Cr in 5% HCl
Hydrochloric acid: 5.0%; Chromium: 0.1%; Water: 94.9%
Clear colorless
Chromium

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-595-7