missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13864AE

 View more versions of this product

Catalog No. AA13864AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Chromium
Clear liquid
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chromium
23976
Liquid
Colorless
7440-47-3,7647-01-0,7732-18-5
Cr
Hydrochloric acid: 5.0%; Chromium: 0.1%; Water: 94.9%
[Cr]
51.996
CHEBI:28073
23 hPa (17mm Hg) at 20°C
100mL

Chemical Identifiers

7440-47-3
51.996
VYZAMTAEIAYCRO-UHFFFAOYSA-N
CHEBI:28073
[Cr]
Cr
MFCD00010944
23976
chromium
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

EINECSNumber : 231-595-7

SDS Documents