Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium, Oil based standard solution, Specpure™, Cr 1000μg/g

Chemical Identifiers

Chemical Name or Material Chromium Oil based standard solution
Name Note Cr 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00010944
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1422218

 
alfa aesar™
1422218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Chromium Oil based standard solution
Quantity 50g
Name Note Cr 1000μg/g
Purity Grade Specpure™
Physical Form Liquid
MDL Number MFCD00010944
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.