missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chromium nitride, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01214914
Description
Chromium nitride is used for stainless steel, corrosion resistance steel, alloy steel and such special steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chromium nitride | |
Powder | |
CrN | |
Insoluble in water. | |
66.00 |
12053-27-9 | |
25 g | |
MFCD00016048 | |
YTZQKGWLQOLFNF-UHFFFAOYSA-N |
Chemical Identifiers
12053-27-9 | |
66.00 | |
YTZQKGWLQOLFNF-UHFFFAOYSA-N |
CrN | |
MFCD00016048 |
Safety and Handling
GHS H Statement
H302-H317
P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
H302-H317
EINECSNumber : 235-002-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only