Please login to your online account to display your discounted pricing

Chromium(III) trifluoride 99%, ACROS Organics™


Quantity 5g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC383740050

 View more versions of this product

Catalog No. AC383740050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5% (ex fluorine)
Formula Weight 108.99
Physical Form Crystals or Crystalline Powder
Quantity 5g
Color Green
Merck Index 15, 2227
Packaging Plastic bottle
CAS 7788-97-8
Chemical Name or Material Chromium(III) trifluoride
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00016045
Linear Formula CrF3
Molecular Formula CrF3
Assay Percent Range 99%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Contact with acids liberates toxic gas.

Specifications

Chemical Name or Material Chromium(III) trifluoride
Assay ≥98.5% (ex fluorine)
Assay Percent Range 99%
Molecular Formula CrF3
Linear Formula CrF3
Formula Weight 108.99
MDL Number MFCD00016045
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep container tightly closed.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Contact with acids liberates toxic gas.

SDS & Safety Documents