Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(III) telluride, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Chromium(III) telluride
CAS 12053-39-3
Formula Weight 486.79
MDL Number MFCD00016040
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4020606

  SDS
alfa aesar™
4020606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4020614

  SDS
alfa aesar™
4020614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 486.79
Quantity 5g
CAS 12053-39-3
Assay Percent Notes (metals basis)
Health Hazard 1 Danger
Mesh Size −60+100 Mesh
Health Hazard 3 GHS P Statement
P261-P280-P281-P301+P310-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Chromium(III) telluride
Color Gray-black
MDL Number MFCD00016040
Physical Form Powder
Assay 99.5%
TSCA TSCA
Health Hazard 2 GHS H Statement
H301-H350-H332-H317
Toxic if swallowed.
May cause cancer.
Harmful if inhaled.
May cause an allergic skin reaction.
Odor Odorless
Density 7.0g/mL
Melting Point ≈1300°C
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P301+P310-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H350-H332-H317
Toxic if swallowed.
May cause cancer.
Harmful if inhaled.
May cause an allergic skin reaction.