Please login to your online account to display your discounted pricing

Chromium(III) standard solution, for AAS, 1 mg/ml Cr in 2% HCl, ACROS Organics™


Quantity 500mL
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  195915000

 View more versions of this product

Catalog No. AC195915000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 52
Quantity 500mL
Color Blue-green
Purity Grade For AAS
Concentration 997 to 1003 μg/mL of Cr(3+) in 2% HCl (20°C)
Density 0.9800g/mL
Chemical Name or Material Chromium(III) standard solution
Name Note 1 mg/ml Cr in 2% HCl
Molecular Formula Cr
CAS 7440-47-3
Health Hazard 1 Warning
CAS 7647-01-0
Health Hazard 3 GHS P Statement:
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle
CAS 7732-18-5
MDL Number MFCD00143970
Health Hazard 2 GHS H Statement:
May be corrosive to metals.

Specifications

Chemical Name or Material Chromium(III) standard solution
Name Note 1 mg/ml Cr in 2% HCl
CAS 7440-47-3
CAS 7647-01-0
CAS 7732-18-5
Purity Grade For AAS
Molecular Formula Cr
Formula Weight 52
MDL Number MFCD00143970
Accessories

Warning

GHS P Statement:
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be corrosive to metals.

SDS & Safety Documents