Please login to your online account to display your discounted pricing

Chromium(III) standard solution, for AAS, 1 mg/ml Cr in 2% HCl, ACROS Organics™

$20.94 - $71.31

Chemical Identifiers

Chemical Name or Material Chromium(III) standard solution
Name Note 1 mg/ml Cr in 2% HCl
CAS 7440-47-3
CAS 7647-01-0
CAS 7732-18-5
Purity Grade For AAS
Molecular Formula Cr
Formula Weight 52
MDL Number MFCD00143970
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC195915000

  SDS
thermo fisher scientific
195915000
Plastic bottle 500mL Each for $71.31

AC195911000

  SDS
fisher scientific acros
195911000
Plastic bottle 100mL Each for $20.94
Description & Specifications

Specifications

Physical Form Solution
Formula Weight 52
Quantity 500mL
Color Blue-green
Purity Grade For AAS
Concentration 997 to 1003 μg/mL of Cr(3+) in 2% HCl (20°C)
Density 0.9800g/mL
CAS 7732-18-5
CAS 7647-01-0
Health Hazard 1 Warning
Molecular Formula Cr
Name Note 1 mg/ml Cr in 2% HCl
Chemical Name or Material Chromium(III) standard solution
CAS 7440-47-3
Health Hazard 3 GHS P Statement:
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
May be corrosive to metals.
MDL Number MFCD00143970

For atomic adsorption spectroscopy

Safety and Handling

Warning

GHS P Statement:
Keep only in original container.
Absorb spillage to prevent material damage.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May be corrosive to metals.