Please login to your online account to display your discounted pricing

Chromium III Speciation Standard (Assurance™ Grade/For ICP-MS), SPEX CertiPrep

$81.14 - $81.14

Chemical Identifiers

Chemical Name or Material Chromium III Speciation Standard
CAS Cr+3
CAS Min % 0.1
CAS Max % 0.1
CAS 7697-37-2
CAS Min % 2.0
CAS Max % 2.0
CAS 7732-18-5
CAS Min % 97.9
CAS Max % 97.9
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPECCR3M

 
spex certiprep llc
SPXSPECCR3M
Triple-leached LDPE bottle 30mL Each for $81.14
Description & Specifications

Specifications

Quantity 30mL
CAS Max % 97.9
Purity Grade Assurance Grade for ICP-MS
CAS Cr+3
CAS Min % 0.1
CAS Max % 0.1
CAS 7697-37-2
CAS Min % 2.0
CAS Max % 2.0
CAS 7732-18-5
CAS Min % 97.9
Chemical Name or Material Chromium III Speciation Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Health Hazard 1 Causes skin irritation, serious eye irritation
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
DOT Information Nonhazardous
Solvent or Matrix 2% HNO3
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Causes skin irritation, serious eye irritation

Ambient