missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium(III) potassium sulfate dodecahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03671522

 View more versions of this product

Catalog No. AA3671522



Description

Description

Chromium(III) potassium sulfate dodecahydrate used for leather tanning, dyeing mordant and photographic fuser agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chromium(III) potassium sulfate dodecahydrate
7788-99-0
Solid
CrH24KO20S2
MFCD00149917
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
ZFVHBEKVAITXHW-UHFFFAOYSA-J
potassium;chromium(3+);disulfate;dodecahydrate
24596
98.0 to 102.0%
Odorless
89°C
Purple
100 g
CrK(SO4)2·12H2O
14,2229
Soluble in water.
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
499.386
499.39 (283.21 Anhydrous)
ACS
1.83 g/mL

Chemical Identifiers

7788-99-0
499.386
ZFVHBEKVAITXHW-UHFFFAOYSA-J
24596
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
CrH24KO20S2
MFCD00149917
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
potassium;chromium(3+);disulfate;dodecahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 233-401-6

RTECSNumber : GB6850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only