missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium(III) potassium sulfate dodecahydrate, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4408322

 View more versions of this product

Catalog No. AA4408322



Specifications

Specifications

Chromium(III) potassium sulfate dodecahydrate
99.9% (Metal basis)
MFCD00149917
acmc-20alga
ZFVHBEKVAITXHW-UHFFFAOYSA-J
potassium;chromium(3+);disulfate;dodecahydrate
24596
Crystalline
Odorless
89°C
7788-99-0
CrH24KO20S2
Violet crystals
Soluble in water 24g/100 mL (15°C).
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
499.386
499.39 (283.21 Anhydrous)
1.83
Violet
100g

Chemical Identifiers

7788-99-0
499.386
ZFVHBEKVAITXHW-UHFFFAOYSA-J
24596
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
CrH24KO20S2
MFCD00149917
acmc-20alga
potassium;chromium(3+);disulfate;dodecahydrate
Safety and Handling

Safety and Handling

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 233-401-6

RTECSNumber : GB6850000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents