Learn More
Chromium(III) phosphate tetrahydrate, Thermo Scientific Chemicals
For Use As a Catalyst, Pigment
Supplier: Thermo Scientific Chemicals 02411009
Description
It is used as an additive in drugs, anti-corrosive pigments, catalysts and polymer manufacture, is also used to catalyze cation exchange in sorption reactions, as a laminated structure to dampen vibration and noise in a motor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chromium(III) phosphate tetrahydrate | |
Powder | |
CrH8O8P | |
chromium iii phosphate tetrahydrate, cr.po4.4h2o, chromium 3+ ion tetrahydrate phosphate, chromium 3+ phosphate-water 1/1/4, chromium 3+ tetrahydrate phosphate | |
BVZHAJJRIRDSTG-UHFFFAOYSA-K | |
chromium(3+);phosphate;tetrahydrate | |
53471880 | |
Dark powder |
10101-59-4 | |
10 g | |
MFCD00016049 | |
Soluble in acids. Insoluble in water. | |
O.O.O.O.[O-]P(=O)([O-])[O-].[Cr+3] | |
219.026 | |
219.04 (146.97 Anhydrous) | |
2.26 g/cm3 |
Chemical Identifiers
10101-59-4 | |
219.026 | |
BVZHAJJRIRDSTG-UHFFFAOYSA-K | |
53471880 | |
O.O.O.O.[O-]P(=O)([O-])[O-].[Cr+3] |
CrH8O8P | |
MFCD00016049 | |
chromium iii phosphate tetrahydrate, cr.po4.4h2o, chromium 3+ ion tetrahydrate phosphate, chromium 3+ phosphate-water 1/1/4, chromium 3+ tetrahydrate phosphate | |
chromium(3+);phosphate;tetrahydrate |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 232-141-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only