missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.8% (metals basis)

$990.30 - $990.30

Chemical Identifiers

CAS 1308-38-9
Formula Weight 151.99
MDL Number MFCD00010949
Chemical Name or Material Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA41059KS SDS Alfa Aesar™
41059KS
1 each Each for $990.30
Specifications Description & Specifications

Chemical Identifiers

1308-38-9
151.99
MFCD00010949
Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick

Specifications

151.99
3000°C
5.22g/mL
Odorless
Pale to dark green
Chromium(III) oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
2435°C
MFCD00010949
1 each
Target
1308-38-9
99.8%, (metals basis)

Safety and Handling

GHS H Statement
H302-H332-H317
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : GB6475000

TSCA : TSCA