Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(III) oxide sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.8% (metals basis)

Magnetron sputtering source

$1,105.20 - $1,105.20

Chemical Identifiers

Chemical Name or Material Chromium(III) oxide sputtering target
CAS 1308-38-9
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Formula Weight 151.99
MDL Number MFCD00010949
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41056KS

 
alfa aesar™
41056KS
1 each Each for $1,105.20
Description & Specifications

Specifications

Formula Weight 151.99
TSCA TSCA
Odor Odorless
Color Pale to dark green
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Chromium(III) oxide sputtering target
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Physical Form Target
Assay 99.8%
Assay Percent Notes (metals basis)
MDL Number MFCD00010949
CAS 1308-38-9
Boiling Point 3000°C
Melting Point 2435°C
Quantity 1 each
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332-H317
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
Density 5.22g/mL
RTECS Number GB6475000
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332-H317
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.