missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(III) oxide, Puratronic™, 99.97% (metals basis)

Manufacturer:  Alfa Aesar™ 1068836

 View more versions of this product

Catalog No. AA1068836



Specifications

Specifications

Chromium(III) oxide
(metals basis)
MFCD00010949
chrome green, chrome ochre, chrome oxide, chromia, chromium 3+ oxide, chromium iii oxide, chromium sesquioxide, dichromium trioxide, green chromium oxide, green cinnabar
QDOXWKRWXJOMAK-UHFFFAOYSA-N
oxo(oxochromiooxy)chromium
517277
151.99
99.97%
5.22 g/mL
Odorless
3000°C
2435°C
1308-38-9
Cr2O3
14,2234
Insoluble in water,alcohol,acetone,alkalies and acids.
O=[Cr]O[Cr]=O
151.989
CHEBI:48242
-22 Mesh Powder
Puratronic™
−22 Mesh
2.551
Green
500g

Chemical Identifiers

1308-38-9
151.989
QDOXWKRWXJOMAK-UHFFFAOYSA-N
517277
oxo(oxochromiooxy)chromium
Cr2O3
MFCD00010949
chrome green, chrome ochre, chrome oxide, chromia, chromium 3+ oxide, chromium iii oxide, chromium sesquioxide, dichromium trioxide, green chromium oxide, green cinnabar
CHEBI:48242
O=[Cr]O[Cr]=O
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H317
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 215-160-9

RTECSNumber : GB6475000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents