missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chromium(III) oxide, 99%, amorphous powder, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 192080025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Chromium(III) oxide | |
2.5kg | |
Cr2O3 | |
03,38; 13,338 | |
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide | |
QDOXWKRWXJOMAK-UHFFFAOYSA-N | |
oxo(oxochromiooxy)chromium | |
517277 | |
151.99 | |
Plastic Bottle | |
5.2100g/mL |
1308-38-9 | |
98.5% min | |
Cr2O3 | |
15,2238 | |
Solubility in water: insoluble in water. Other solubilities: slightly soluble in acids and alkalies,practically insoluble in alcohol and acetone | |
O=[Cr]O[Cr]=O | |
151.99 | |
CHEBI:48242 | |
99% | |
5.21 |
Safety and Handling
EINECSNumber : 215-160-9
RUO â Research Use Only