missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium(III) oxide, 99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1228536

 View more versions of this product

Catalog No. AA1228536



Specifications

Specifications

Chromium(III) oxide
99%
MFCD00010949
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
O=[Cr]O[Cr]=O
151.989
CHEBI:48242
Powder
Green
1308-38-9
Cr2O3
Pale to dark green powder
QDOXWKRWXJOMAK-UHFFFAOYSA-N
oxo(oxochromiooxy)chromium
517277
151.99
Odorless
500g

Chemical Identifiers

1308-38-9
151.989
QDOXWKRWXJOMAK-UHFFFAOYSA-N
517277
oxo(oxochromiooxy)chromium
Cr2O3
MFCD00010949
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
CHEBI:48242
O=[Cr]O[Cr]=O
Safety and Handling

Safety and Handling

H302-H332-H317 : Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction.

EINECSNumber : 215-160-9

Documents