Learn More
Chromium(III) oxide, 99.6% (metals basis), Thermo Scientific Chemicals
$94.89 - $352.86
Chemical Identifiers
CAS | 1308-38-9 |
---|---|
Molecular Formula | Cr2O3 |
Molecular Weight (g/mol) | 151.99 |
MDL Number | MFCD00010949 |
InChI Key | UOUJSJZBMCDAEU-UHFFFAOYSA-N |
Synonym | chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide |
PubChem CID | 517277 |
ChEBI | CHEBI:48242 |
IUPAC Name | dichromium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Cr+3].[Cr+3] |
Description
For vacuum deposition. It can be used in selective oxidation. It is mainly used for smelting metal chromium and chromium carbide. It is used as coloring agent for color painting on enamelware and porcelain ware, imitation leather or building material. It is also used for making sun-proof coating, abrasive, green polishing compound and special printing ink for printing bank note. It is used as catalyst for organic synthetic chromium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1308-38-9 | |
151.99 | |
UOUJSJZBMCDAEU-UHFFFAOYSA-N | |
517277 | |
dichromium(3+) trioxidandiide |
Cr2O3 | |
MFCD00010949 | |
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide | |
CHEBI:48242 | |
[O--].[O--].[O--].[Cr+3].[Cr+3] |
Specifications
1308-38-9 | |
25 g | |
Cr2O3 | |
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide | |
[O--].[O--].[O--].[Cr+3].[Cr+3] | |
151.99 | |
CHEBI:48242 | |
5.22g/mL |
Fused Lump | |
99.6% (metals basis) | |
MFCD00010949 | |
UOUJSJZBMCDAEU-UHFFFAOYSA-N | |
dichromium(3+) trioxidandiide | |
517277 | |
151.99 | |
Chromium oxide |
Safety and Handling
EINECSNumber : 215-160-9
RUO – Research Use Only