missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium(III) nitrate nonahydrate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 219200050

 View more versions of this product

Catalog No. AC219200050


 

Specifications Description & Specifications

Specifications

≥98.5%
Crystalline Chunks
Dark purple to black
2150954
50ppm max.
0.2% max. (as SO4)
50ppm max.
MFCD00149659
20ppm max.
10ppm max.
10ppm max.
Glass bottle
50ppm max.
10ppm max.
99%
Chromium(III) nitrate nonahydrate
400.14
5g
2 to 3 (5% aq. soln.)
50ppm max.
50ppm max.
10ppm max.
CrN3O9·9H2O
99%
20ppm max.
10ppm max.
Chromic nitrate
50ppm max.
200ppm max.
10ppm max.
Cr(NO3)3·9H2O

Chemical Identifiers

2150954
CrN3O9·9H2O
400.14
Chromic nitrate
≥98.5%
99%
Cr(NO3)3·9H2O
MFCD00149659
Chromium(III) nitrate nonahydrate, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.

GHS P Statement:
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

RTECSNumber : GB6300000