Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Chromium(III) nitrate hydrate
CAS 20249-21-2
Purity Grade Puratronic™
Linear Formula Cr(NO3)3·xH2O
Formula Weight 238.01g/mol (Anhydrous)
MDL Number MFCD00010946
UN Number UN2720
Merck Index 14,2226
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1300506

 
alfa aesar™
13005-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Color Dark purple
Melting Point 60°C
Quantity 5g
Chemical Name or Material Chromium(III) nitrate hydrate
CAS 20249-21-2
Purity Grade Puratronic™
Physical Form Crystalline
Assay 99.999%
Assay Percent Notes (metals basis)
Linear Formula Cr(NO3)3·xH2O
Formula Weight 238.01g/mol (Anhydrous)
MDL Number MFCD00010946
UN Number UN2720
Merck Index 14,2226
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H272-H301-H332-H319
May intensify fire
oxidizer.
Toxic if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P221-P210-P301+P310-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: CHROMIUM NITRATE
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P221-P210-P301+P310-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H272-H301-H332-H319
May intensify fire
oxidizer.
Toxic if swallowed.
Harmful if inhaled.
Causes serious eye irritation.