Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(III) hexafluoro-2,4-pentanedionate

Chemical Identifiers

Chemical Name or Material Chromium(III) hexafluoro-2,4-pentanedionate
CAS 14592-80-4
Formula Weight 673.14
MDL Number MFCD00044995
Synonym Chromium(III) hexafluoroacetylacetonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3858103

  SDS
alfa aesar™
3858103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3858106

  SDS
alfa aesar™
3858106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 673.14
Chemical Name or Material Chromium(III) hexafluoro-2,4-pentanedionate
Odor Odorless
Physical Form Crystalline
MDL Number MFCD00044995
CAS 14592-80-4
Synonym Chromium(III) hexafluoroacetylacetonate
Color Green
Melting Point 83° to 88°C
Quantity 1g
Solubility Information Insoluble in water
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.