missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(III) chloride hexahydrate, Puratronic™, 99.998% (metals basis)

Manufacturer:  Alfa Aesar™ 1068614

 View more versions of this product

Catalog No. AA1068614



Specifications

Specifications

Chromium(III) chloride hexahydrate
(metals basis)
CrCl3·6H2O
UN3260
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
O.O.O.O.O.O.Cl[Cr](Cl)Cl
266.436
CHEBI:53442
Crystalline Aggregates
Puratronic™
Odorless
83°C
10060-12-5
Cl3CrH12O6
MFCD00149660
14,2222
LJAOOBNHPFKCDR-UHFFFAOYSA-K
trichlorochromium;hexahydrate
104957
266.45 (158.35 Anhydrous)
99.998%
1.835g/mL
Green
25g

Chemical Identifiers

10060-12-5
266.436
LJAOOBNHPFKCDR-UHFFFAOYSA-K
104957
trichlorochromium;hexahydrate
Cl3CrH12O6
MFCD00149660
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech
CHEBI:53442
O.O.O.O.O.O.Cl[Cr](Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H334-H314-H318-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : GB5450000

TSCA : TSCA

Documents