Learn More
Chromium(III) chloride hexahydrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 213360051
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chromium(III) chloride hexahydrate | |
10060-12-5 | |
Crystalline Powder | |
98% | |
[Cr(H2O)4Cl2]Cl·2H2O | |
14, 2222 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,slightly soluble in acetone,practically insoluble in ether | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] | |
266.44 | |
CHEBI:53442 | |
98% |
83.0°C | |
Green | |
5 kg | |
Cl3CrH12O6 | |
MFCD00149660 | |
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech | |
LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
chromium(3+) hexahydrate trichloride | |
104957 | |
266.44 | |
Plastic drum |
Chemical Identifiers
10060-12-5 | |
266.44 | |
LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
104957 | |
chromium(3+) hexahydrate trichloride |
Cl3CrH12O6 | |
MFCD00149660 | |
chromium iii chloride hexahydrate, chromic chloride hexahydrate, hexaaquachromium chloride, chromium trichloride hexahydrate, trichlorochromium hexahydrate, chromium chloride hexahydrate, hexaaquachromium iii chloride, chrom iii-chlorid, chromium 3+ chloride hexahydrate, chlorid chromity hexahydrat czech | |
CHEBI:53442 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May be corrosive to metals.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Absorb spillage to prevent material damage.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SK
GHS Signal Word: Warning
RUO – Research Use Only