Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(III) benzoylacetonate, 98%

$69.27 - $250.91

Chemical Identifiers

Chemical Name or Material Chromium(III) benzoylacetonate
CAS 16432-36-3
Molecular Formula C30H27CrO6
Formula Weight 535.54
MDL Number MFCD00036117
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1605103

 
alfa aesar™
A1605103
1g Each for $69.27

AAA1605106

 
alfa aesar™
A1605106
5g Each for $250.91
Description & Specifications

Specifications

Formula Weight 535.54
Chemical Name or Material Chromium(III) benzoylacetonate
TSCA TSCA
Assay 98%
Molecular Formula C30H27CrO6
MDL Number MFCD00036117
CAS 16432-36-3
Quantity 1g
RTECS Number GB7580000