Please login to your online account to display your discounted pricing

Chromium(III) acetylacetonate, 97%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  192131000

 View more versions of this product

Catalog No. AC192131000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder, Crystals, and/or Chunks
Formula Weight 349.32
Quantity 100g
Color Purple to maroon
Assay 14.4 to 15.3% (Cr)
Infrared Spectrum Authentic
Chemical Name or Material Chromium(III) acetylacetonate
Assay Percent Range 97%
Packaging Plastic bottle
Molecular Formula C15H21CrO6
Linear Formula (CH3COCH=C(CH3)O)3Cr
MDL Number MFCD00000015
CAS 21679-31-2
Health Hazard 1 Warning
Synonym Chromium(III) 2, 4-pentanedionate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Chromium(III) acetylacetonate
Assay 14.4 to 15.3% (Cr)
Assay Percent Range 97%
Molecular Formula C15H21CrO6
Linear Formula (CH3COCH=C(CH3)O)3Cr
Formula Weight 349.32
MDL Number MFCD00000015
Synonym Chromium(III) 2, 4-pentanedionate
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents