Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(III) 2-ethylhexanoate, 50% in 2-ethylhexanoic acid

$113.62 - $426.42

Chemical Identifiers

Chemical Name or Material Chromium(III) 2-ethylhexanoate
CAS 3444-17-5
Molecular Formula C24H45CrO6
Formula Weight 481.62
MDL Number MFCD00014005
Synonym 2-Ethylhexanoic acid chromium(III) salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1656018

 
alfa aesar™
A1656018
50g Each for $113.62

AAA1656030

 
alfa aesar™
A1656030
250g Each for $426.42
Description & Specifications

Specifications

Formula Weight 481.62
CAS 3444-17-5
TSCA TSCA
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Chemical Name or Material Chromium(III) 2-ethylhexanoate
Health Hazard 2 GHS H Statement
H361-H315-H319
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
Concentration 50% in 2-ethylhexanoic acid
Health Hazard 1 Warning
Molecular Formula C24H45CrO6
Quantity 50g
MDL Number MFCD00014005
Synonym 2-Ethylhexanoic acid chromium(III) salt
Odor Weak, Characteristic
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H361-H315-H319
Suspected of damaging fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.