missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(III) 2,4-pentanedionate, 97%

Catalyst in the oxidation of methacrylic esters by hydrogen peroxide, MOCVD

Manufacturer:  Alfa Aesar™ 1253836

 View more versions of this product

Catalog No. AA1253836



Specifications

Specifications

Chromium(III) 2,4-pentanedionate
340°C
500g
C15H24CrO6
chromium iii acetylacetonate, dsstox_cid_30421, dsstox_gsid_51863
MJSNUBOCVAKFIJ-LNTINUHCSA-N
chromium;(Z)-4-oxoniumylidenepent-2-en-2-olate
91759531
Crystalline
1.35g/mL
216°C
21679-31-2
MFCD00000015
Insoluble in water; Soluble in toluene, acetic acid
CC(=CC(=[OH+])C)[O-].CC(=CC(=[OH+])C)[O-].CC(=CC(=[OH+])C)[O-].[Cr]
352.347
349.33
97%

Chemical Identifiers

21679-31-2
352.347
MJSNUBOCVAKFIJ-LNTINUHCSA-N
91759531
CC(=CC(=[OH+])C)[O-].CC(=CC(=[OH+])C)[O-].CC(=CC(=[OH+])C)[O-].[Cr]
C15H24CrO6
MFCD00000015
chromium iii acetylacetonate, dsstox_cid_30421, dsstox_gsid_51863
chromium;(Z)-4-oxoniumylidenepent-2-en-2-olate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : GB7575000

TSCA : TSCA

Documents