Please login to your online account to display your discounted pricing

Chromium(II) chloride, 99.9%, (trace metal basis), ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC315190250

 View more versions of this product

Catalog No. AC315190250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Physical Form Powder
Formula Weight 122.9
Quantity 25g
Color Off-white to green-gray
Fieser 01,149; 02,76; 03,60; 04,144; 07,73; 09,511; 12,136; 13,84; 14,94; 16,93; 17,84
Merck Index 15, 2248
Linear Formula CrCl2
Total Trace Metal Impurities 0.1% max.
Chemical Name or Material Chromium(II) chloride
Assay Percent Range 99.9%
Molecular Formula Cl2Cr
CAS 10049-05-5
Packaging Glass bottle
MDL Number MFCD00010947
Synonym Chromous chloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Chromium(II) chloride
CAS 10049-05-5
Purity Grade Trace Metal Basis
Assay Percent Range 99.9%
Molecular Formula Cl2Cr
Linear Formula CrCl2
Formula Weight 122.9
MDL Number MFCD00010947
Synonym Chromous chloride
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents