missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(II) chloride, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4717603

 View more versions of this product

Catalog No. AA4717603



Specifications

Specifications

Chromium(II) chloride
(metals basis)
MFCD00010947
14,2244
122.9
99.9%
−80 Mesh
Air & Moisture Sensitive
824°C
10049-05-5
CrCl2
UN3260
Chromous chloride
Powder
2.878g/mL
Odorless
Gray
1g

Chemical Identifiers

10049-05-5
MFCD00010947
Chromous chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : GB5250000

TSCA : TSCA

SDS Documents