Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium(II) chloride, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Chromium(II) chloride
CAS 10049-05-5
Linear Formula CrCl2
Formula Weight 122.9
MDL Number MFCD00010947
UN Number UN3260
Merck Index 14,2244
Synonym Chromous chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4717603

  SDS
alfa aesar™
4717603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4717606

  SDS
alfa aesar™
4717606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4717614

  SDS
alfa aesar™
4717614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.9
Chemical Name or Material Chromium(II) chloride
TSCA TSCA
Mesh Size −80 Mesh
Odor Odorless
Physical Form Powder
Assay 99.9%
Sensitivity Air & Moisture Sensitive
Assay Percent Notes (metals basis)
Linear Formula CrCl2
MDL Number MFCD00010947
CAS 10049-05-5
UN Number UN3260
Merck Index 14,2244
Synonym Chromous chloride
Color Gray
Melting Point 824°C
Quantity 1g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 2.878g/mL
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
RTECS Number GB5250000
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.