missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium chunks, 2-15mm (0.08-0.59in), 99.996% (metals basis)

Manufacturer:  Alfa Aesar™ 4456518

 View more versions of this product

Catalog No. AA4456518



Specifications

Specifications

Chromium chunks
2 to 15mm (0.08 to 0.59 in.)
(metals basis)
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
[Cr]
51.996
CHEBI:28073
50g
7440-47-3
Cr
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
Odorless
chromium
23976
99.996%

Chemical Identifiers

7440-47-3
51.996
VYZAMTAEIAYCRO-UHFFFAOYSA-N
23976
chromium
Cr
MFCD00010944
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
CHEBI:28073
[Cr]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents