Please login to your online account to display your discounted pricing

Chromium Chloride, X-Hydrate, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$886.27 - $2,138.51

Chemical Identifiers

Chemical Name or Material Chromium Chloride
CAS 10060-12-5
Name Note X-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula CrCl3·6H2O
Formula Weight 266.45
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002599

 
avantor performance
158805
Poly Bottle 2.5kg Each for $2,138.51

02002600

 
avantor performance
158801
Poly Bottle 500g Each for $886.27
Description & Specifications

Specifications

Formula Weight 266.45
Chemical Name or Material Chromium Chloride
Molecular Formula CrCl3·6H2O
CAS 10060-12-5
Name Note X-Hydrate
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 1.76kg
Quantity 2.5kg

    For Laboratory,Research,or Manufacturing Use