Please login to your online account to display your discounted pricing

Chromium carbide, 99.5% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Chromium carbide
CAS 12105-81-6
Physical Form Powder
Assay Percent Range 99.5% (Metal basis)
Molecular Formula Cr23C6
Formula Weight 1267.97
MDL Number MFCD00678358
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4231309

 
alfa aesar
4231309
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4231318

 
alfa aesar
4231318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4231330

 
alfa aesar
4231330
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1267.97
Quantity 10g
Color Gray
DOT Information DOT Class: 4.1, Packaging Group: III
Chemical Name or Material Chromium carbide
EINECS Number 235-160-2
Odor Odorless
Physical Form Powder
Assay Percent Range 99.5% (Metal basis)
Molecular Formula Cr23C6
Appearance Gray powder
MDL Number MFCD00678358
CAS 12105-81-6
Health Hazard 1 H228; Flammable solid.
Safety and Handling

H228; Flammable solid.