Please login to your online account to display your discounted pricing

Alfa Aesar™ Chromium carbide, 99.5% (metals basis)

Chemical Identifiers

Chemical Name or Material Chromium carbide
CAS 12012-35-0
Formula Weight 180.02
MDL Number MFCD00016041
UN Number UN3178
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1154718

  SDS
alfa aesar™
1154718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1154730

  SDS
alfa aesar™
1154730
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.02
Assay Percent Notes (metals basis)
Mesh Size −325 Mesh
CAS 12012-35-0
Health Hazard 2 GHS H Statement
H228
Flammable solid.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.
Melting Point 1890°C
Chemical Name or Material Chromium carbide
MDL Number MFCD00016041
Physical Form Powder
Odor Odorless
UN Number UN3178
Quantity 50g
Health Hazard 1 Warning
TSCA TSCA
Assay 99.5%
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

GHS H Statement
H228
Flammable solid.