Please login to your online account to display your discounted pricing

Chromium boride, Gray powder, 99% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Chromium boride
CAS 12007-16-8
Physical Form Powder
Assay Percent Range 99% (Metals basis)
Molecular Formula CrB2
Formula Weight 73.62
MDL Number MFCD00016043
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1257014

 
alfa aesar
1257014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1257022

 
alfa aesar
1257022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 73.62
Quantity 25g
Color Gray
Chemical Name or Material Chromium boride
Odor Odorless
Assay Percent Range 99% (Metals basis)
Molecular Formula CrB2
MDL Number MFCD00016043
CAS 12007-16-8