missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromium, AAS standard solution, Specpure™, Cr 1000μg/mL, Alfa Aesar™

Manufacturer:  Alfa Aesar 88059AP

 View more versions of this product

Catalog No. AA88059AP



Specifications

Specifications

Chromium
7789-02-8,7697-37-2,7732-18-5
MFCD00010944
PHFQLYPOURZARY-UHFFFAOYSA-N
chromium(3+);trinitrate;nonahydrate
9887206
17mm Hg at 20°C
500mL
AAS Standard Solution
CrN3O9
Nitric acid: 5%; Chromium(III) nitrate nonahydrate: 0.77%; Water: 94.23%
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
238.01
Liquid
Colorless

Chemical Identifiers

7789-02-8
238.01
PHFQLYPOURZARY-UHFFFAOYSA-N
chromium(3+);trinitrate;nonahydrate
CrN3O9
MFCD00010944
9887206
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]
Safety and Handling

Safety and Handling

When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 231-595-7

SDS Documents