Please login to your online account to display your discounted pricing

Chromic Acid, Reagent Grade, LabChem™

$223.51 - $223.51

Chemical Identifiers

Chemical Name or Material Chromic Acid
CAS 1333-82-0
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula CrO3
Formula Weight 99.99
UN Number UN1463
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC130901

 
lab chem
LC130901
500g Each for $223.51
Description & Specifications

Specifications

Formula Weight 99.99
Chemical Name or Material Chromic Acid
EINECS Number 215-607-8
Solubility Soluble in water
Specific Gravity 2700kg/m3
Storage Information Room Temperature
Melting Point 196°C
Purity Grade Reagent Grade
Physical Form Crystals
Linear Formula CrO3
CAS 1333-82-0
UN Number UN1463
Density 2,700kg/m3
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
May cause fire or explosion; strong oxidizer.
Fatal if inhaled.
Toxic if swallowed or in contact with skin.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames.
No smoking.
Keep away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
Wear protective gloves, flame resistant protective clothing, eye protection, face protection.
Wear respiratory protection.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin, hair or clothing: Rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the risk of explosion.
Collect spillage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
Identification Passes Test
Packaging Poly Bottle
Color Dark red
Quantity 500g
Safety and Handling

Danger

GHS P Statement
Do not handle until all safety precautions have been read and understood.
Keep away from heat, sparks, open flames.
No smoking.
Keep away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
Wear protective gloves, flame resistant protective clothing, eye protection, face protection.
Wear respiratory protection.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin, hair or clothing: Rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
In case of fire: Use carbon dioxide, powder, alcohol-resistant foam to extinguish.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the risk of explosion.
Collect spillage.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May cause fire or explosion; strong oxidizer.
Fatal if inhaled.
Toxic if swallowed or in contact with skin.
Causes severe skin burns and eye damage.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
May cause genetic defects.
May cause cancer.
Suspected of damaging fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.