missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromic Acid, Certified, 10.0% (w/v) ±0.5%, LabChem™

Manufacturer:  LabChem LC131004

 View more versions of this product

Catalog No. LC131004



Specifications

Specifications

Chromic Acid
4L
90,10
CrO3
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
O=[Cr](=O)=O
99.993
CHEBI:48240
Liquid
Certified
Poly Bottle
Room Temperature
Oxygen; Thermal decomposition generates: Corrosive vapors
Orange
1333-82-0,7732-18-5
CrO3
UN1755
WGLPBDUCMAPZCE-UHFFFAOYSA-N
trioxochromium
14915
99.99
10.0% ± 0.5% (w/v)
Passes Test
Soluble in water
Traceable to NIST

Chemical Identifiers

1333-82-0
99.993
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
CHEBI:48240
O=[Cr](=O)=O
CrO3
WGLPBDUCMAPZCE-UHFFFAOYSA-N
14915
trioxochromium
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed or if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhal

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face p

Danger

Documents
Promotions

Promotions