Arcor Electronics Chromel™ Wire
Provides very efficient heating
Manufacturer:  Arcor Electronics HOS26GAUGE
 View more versions of this product
Catalog No. 15-540-2E
Description
Description
Provides very efficient heating
Order Info
Supplied in 0.25 lb. (113g) spools. (Feet per spool varies; approximate values are given in chart.)
Specifications
Specifications
0.015 in.
Used to make inoculating needles
1380 Ft./lb.
2150°F
20% Chromium, 80% Nickel
0.40mm
26
8.43 Ohm/m
1177°C
Chromel-A Wire
Documents