missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chrome Yellow, TCI America™

$71.81 - $360.56

Chemical Identifiers

CAS 6054-97-3
Molecular Formula C17H12N2Na2O6S
Molecular Weight (g/mol) 418.331
MDL Number MFCD00021624
InChI Key PYJCZHNASHXDRA-SJNXHXLPSA-N
Synonym Mordant Yellow 3, Sunchromine Yellow MD
PubChem CID 131855863
IUPAC Name (3Z)-6-oxo-3-[(6-sulfonaphthalen-2-yl)hydrazinylidene]cyclohexa-1,4-diene-1-carboxylic acid;sodium
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C=C1NN=C3C=CC(=O)C(=C3)C(=O)O.[Na].[Na]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C054525G View Documents TCI America
C054525G
25g Each for $71.81
 
C0545500G View Documents TCI America
C0545500G
500g Each for $360.56
 
Specifications

Chemical Identifiers

6054-97-3
418.331
PYJCZHNASHXDRA-SJNXHXLPSA-N
131855863
C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C=C1NN=C3C=CC(=O)C(=C3)C(=O)O.[Na].[Na]
C17H12N2Na2O6S
MFCD00021624
Mordant Yellow 3, Sunchromine Yellow MD
(3Z)-6-oxo-3-[(6-sulfonaphthalen-2-yl)hydrazinylidene]cyclohexa-1,4-diene-1-carboxylic acid;sodium

Specifications

Undesignated
6054-97-3
MFCD00021624
PYJCZHNASHXDRA-SJNXHXLPSA-N
(3Z)-6-oxo-3-[(6-sulfonaphthalen-2-yl)hydrazinylidene]cyclohexa-1,4-diene-1-carboxylic acid;sodium
131855863
Crystal-Powder at 20°C
Chrome Yellow
25g
C17H12N2Na2O6S
Mordant Yellow 3, Sunchromine Yellow MD
C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C=C1NN=C3C=CC(=O)C(=C3)C(=O)O.[Na].[Na]
418.331
416.31
≥95.0% (HPLC)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-2045

TSCA : Yes