missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chrome Azurol S, MP Biomedicals™

Chrome Azurol S is used as a reagent for determination of fluoride

$53.14 - $53.14

Chemical Identifiers

CAS 1667-99-8
Molecular Formula C23H13Cl2Na3O9S
Molecular Weight (g/mol) 605.277
MDL Number MFCD00001615
InChI Key FUIZKNBTOOKONL-UHFFFAOYSA-K
Synonym C.I. 43825, Mordant Blue 29
PubChem CID 54736314
IUPAC Name trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate
SMILES CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15498210
View Documents
Mp Biomedicals
0215498210
10g Each for $53.14
 
Specifications

Chemical Identifiers

1667-99-8
605.277
FUIZKNBTOOKONL-UHFFFAOYSA-K
54736314
CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]
C23H13Cl2Na3O9S
MFCD00001615
C.I. 43825, Mordant Blue 29
trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate

Specifications

1667-99-8
MFCD00001615
FUIZKNBTOOKONL-UHFFFAOYSA-K
trisodium;5-[(3-carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)-(2,6-dichloro-3-sulfonatophenyl)methyl]-3-methyl-2-oxidobenzoate
54736314
Powder
Brown
Chrome Azurol S, Dye content: approx. 65%
C23H13Cl2Na3O9S
C.I. 43825, Mordant Blue 29
CC1=CC(=CC(=C1[O-])C(=O)[O-])C(=C2C=C(C(=O)C(=C2)C(=O)O)C)C3=C(C=CC(=C3Cl)S(=O)(=O)[O-])Cl.[Na+].[Na+].[Na+]
605.277
605.29
Chromeazurol
10g
Safety and Handling

Safety and Handling

EINECSNumber : 216-787-0

Recommended Storage : Store at Room Temperature (15-30°C).