Please login to your online account to display your discounted pricing

Chrome Azurol S, ACROS Organics™

$108.34 - $375.54

Chemical Identifiers

Chemical Name or Material Chrome Azurol S
CAS 1667-99-8
Molecular Formula C23H13Cl2Na3O9S
Formula Weight 605.29
MDL Number MFCD00001615
Synonym C.I. 43825
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC190051000

  SDS
thermo fisher scientific
AC190051000
Glass bottle 100g Each for $375.54

AC190050250

  SDS
thermo fisher scientific
190050250
Glass bottle 25g Each for $108.34
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 605.29
Quantity 100g
Color Dark brown to black
EINECS Number 216-787-0
TSCA TSCA
Synonym C.I. 43825
Infrared Spectrum Authentic
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Molecular Formula C23H13Cl2Na3O9S
CAS 1667-99-8
Chemical Name or Material Chrome Azurol S
Packaging Glass bottle
UV Spectrometry Complies
Lambda Max 424 to 436nm (c=0.03g/L in H2O)
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin
Health Hazard 1 Warning
MDL Number MFCD00001615

Indicator

Safety and Handling

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsin

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.