missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chromate, Ion Chromatography Standard Solution, Alfa Aesar™, Specpure™

Chemical Identifiers

Physical Form Liquid
Molecular Formula K2CrO4 in H2O
MDL Number MFCD00011368
Chemical Name or Material Chromate, CrO4?2 1000μg/mL
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42564AE SDS Alfa Aesar
42564AE
100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA42564AP SDS Alfa Aesar
42564AP
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

Liquid
K2CrO4 in H2O
MFCD00011368
Chromate, CrO4?2 1000μg/mL

Specifications

Clear colorless
100mL
1g/cm3 at 20°C
Clear liquid
Chromate, CrO4-2 1000μg/mL
Liquid
K2CrO4 in H2O
Odorless
MFCD00011368

Safety and Handling

H340-H350: May cause genetic defects. May cause cancer.

EINECSNumber : 232-140-5