Please login to your online account to display your discounted pricing

Chromanol 293B, Tocris

IKs blocker. Also blocks ICFTR

$239.63 - $970.04

Chemical Identifiers

Chemical Name or Material Chromanol 293B
Assay Percent Range >99%
Molecular Formula C15H20N2O4S
Formula Weight 324.39
Synonym trans-N-[6-Cyano-3, 4-dihydro-3-hydroxy-2, 2-dimethyl-2H-1-benzopyran-4-yl]-N-methyl-ethanesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

141250

 
r & d systems
RDS141250
50mg Each for $970.04

141210

 
r & d systems
RDS141210
10mg Each for $239.63
Description & Specifications

Specifications

Formula Weight 324.39
Quantity 50mg
Assay Percent Range >99%
Synonym trans-N-[6-Cyano-3, 4-dihydro-3-hydroxy-2, 2-dimethyl-2H-1-benzopyran-4-yl]-N-methyl-ethanesulfonamide
Chemical Name or Material Chromanol 293B
CAS 163163-23-3
Recommended Storage Store at Room Temperature
Molecular Formula C15H20N2O4S
Safety and Handling

Store at Room Temperature