Please login to your online account to display your discounted pricing

(Chroman-8-ylmethyl)amine, 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (Chroman-8-ylmethyl)amine
CAS 933727-40-3
CAS Min % 88.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 90%
Molecular Formula C10H13NO
Formula Weight 163.22
MDL Number MFCD17171385
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC432660050

  SDS
thermo fisher scientific
AC432660050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥94.0%
Formula Weight 163.22
Quantity 5g
Color Clear pale yellow
CAS Min % 88.0
CAS Max % 100.0
Packaging Glass bottle
Molecular Formula C10H13NO
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Chemical Name or Material (Chroman-8-ylmethyl)amine
Assay Percent Range 90%
MDL Number MFCD17171385
CAS 933727-40-3
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.