Please login to your online account to display your discounted pricing

chroman-8-sulfonyl chloride, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC76103DA

 View more versions of this product

Catalog No. CC76103DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 232.68
Quantity 1g
Assay 97%
Chemical Name or Material chroman-8-sulfonyl chloride
Packaging Amber Glass Bottle
Molecular Formula C9H9ClO3S
CAS 104897015
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material chroman-8-sulfonyl chloride
Assay 97%
Molecular Formula C9H9ClO3S
Formula Weight 232.68
Accessories

GHS Signal Word: Danger

SDS & Safety Documents